Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human CXCL1 (GRO alpha) Recombinant Protein, PeproTech®

Catalog No. 30011500UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
100 μg
25 μg
250 μg
5 μg
500 μg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
30011500UG 500 μg
30011100UG 100 μg
300-11-1MG 1 mg
30011250UG 250 μg
300-11-25UG 25 μg
300-11-5UG 5 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 30011500UG Supplier Gibco™ Supplier No. 30011500UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Recombinant Human GRO-alpha/MGSA is a 7.8 kDa protein consisting of 73 amino acids, including the 'ELR' motif common to the CXC chemokine family that binds to CXCR1 or CXCR2. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

CXCL1, CXCL2, and CXCL3, also known respectively as GRO alpha, GRO beta (MIP-2 alpha) and GRO gamma (MIP-2 beta), are members of the CXC subfamily of chemokines. Mature CXCL1/2/3 proteins bind with high affinity to the IL-8 receptor type B and are potent neutrophil attractants and activators. In vitro, the processed forms GRO-alpha(4-73), GRO-alpha(5-73) and GRO-alpha(6-73) show a 30-fold higher chemotactic activity.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P09341
For Use With (Application) ELISA, Functional Assay, Western Blot
Formulation protein with no preservative
Gene ID (Entrez) 2919
Molecular Weight (g/mol) 7.8 kDa
Name Human CXCL1 (GRO alpha)
Quantity 500 μg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias alpha-chemokine; C Cmotif chemokine; C x C motif chemokine; CC motif chemokine; CCmotif chemokine; chemokine (C-X-C motif) ligand 1; chemokine (C-X-C motif) ligand 1 (melanoma growth stimulating activity, alpha); Cinc1; CINC-1; CXC; CXC motif chemokine; C-X-C motif chemokine 1; C-X-C motif chemokine ligand 1; CXCL; CXCL1; CXCL-1; Cytokine-induced neutrophil chemoattractant 1; fibroblast secretory protein; Fsp; Gro; GRO a; GRO a; Gro1; GRO1 oncogene; GRO1 oncogene (melanoma growth stimulating activity, alpha); GRO1 oncogene (melanoma growth-stimulating activity); GROa; GROalpha; GRO-alpha(1-73); GRO-alpha(4-73); GRO-alpha(5-73); GRO-alpha(6-73); Growth-regulated alpha protein; GROa; Hematopoietic synergistic factor; HSF; KC; KC(5-72); KC/GR)-alpha; KC/GRO-alpha; KC-T; melanoma growth stimulating activity, alpha; Melanoma growth stimulatory activity; melanoma growth stimulatory activity alpha; MGSA; MGSA alpha; MGSA-a; N51; NAP3; NAP-3; Neutrophil-activating protein 3; platelet-derived growth factor-inducible protein KC; SCYB1; secretory protein N51
Common Name CXCL1 (GRO alpha)
Gene Symbol Cxcl1
Biological Activity Determined by its ability to chemoattract human neutrophils using a concentration range of 10.0-50.0 ng/ml
Conjugate Unconjugated
Recombinant Recombinant
Sequence ASVATELRCQ CLQTLQGIHP KNIQSVNVKS PGPHCAQTEV IATLKNGRKA CLNPASPIVK KIIEKMLNSD KSN
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.