Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human CXCL10 (IP-10) Recombinant Protein, PeproTech®

Catalog No. 3001225UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
100 μg
25 μg
250 μg
5 μg
50 μg
500 μg
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
300-12-25UG 25 μg
30012100UG 100 μg
300-12-1MG 1 mg
30012250UG 250 μg
30012500UG 500 μg
300-12-50UG 50 μg
300-12-5UG 5 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Catalog No. 300-12-25UG Supplier Gibco™ Supplier No. 3001225UG
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Recombinant Human IP-10 is an 8.6 kDa protein consisting of 77 amino acids, including the four conserved cysteine residues present in CXC chemokines. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

IP-10 (interferon-gamma induced protein 10 kD; in mouse, also called cytokine responsive gene 2, Crg-2, or mob-1) is a highly inducible, primary response gene that belongs to the CXC chemokine superfamily. It was first cloned in 1985. The biological functions of IP-10 are still unclear. Like Mig, IP-10 has no activity on neutrophils. Its functions include stimulation of monocytes, natural killer and T-cell migration, regulation of T-cell and bone marrow progenitor maturation, modulation of adhesion molecule expression as well as inhibition of angiogenesis. Mig and IP-10 share the same receptor, CXCR3.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P02778
For Use With (Application) ELISA, Functional Assay, Western Blot
Formulation protein with no preservative
Gene ID (Entrez) 3627
Molecular Weight (g/mol) 8.6 kDa
Name Human CXCL10 (IP-10)
Quantity 25 μg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias 10 kDa interferon gamma-induced protein; 10 kDa interferon-gamma induced protein precursor; C Cmotif chemokine; C x C motif chemokine; C7; CC motif chemokine; CCmotif chemokine; chemokine (C-X-C motif) ligand 10; chemokine C-X-C motif ligand 10; chemokine CXC-like protein; Crg2; CRG-2; CXC; CXC chemokine; CXC motif chemokine; C-X-C motif chemokine 10; C-X-C motif chemokine 10-like protein; C-X-C motif chemokine ligand 10; CXCL; CXCL10; CXCL10(1-73); gamma interferon inducible protein 10; gamma IP10; gamma-IP10; gIP-10; H-IP-10; IFI10; Inp10; interferon activated gene 10; interferon-gamma induced protein CRG-2; interferon-gamma-induced protein CRG-2; interferon-gamma-inducible protein 10; interferon-gamma-inducible protein-10; interferon-inducible cytokine IP-10; interferon-inducible protein 10; IP10; IP-10; M-IP-10; Mob1; mob-1; protein 10 from interferon (gamma)-induced cell line; protein Mob-1; Scyb10; similar to Small inducible cytokine B10 precursor (CXCL10) (10 kDa interferon-gamma induced protein) (Gamma-IP10) (IP-10); small inducible cytokine B subfamily (Cys-X-Cys) member 10; small inducible cytokine B subfamily (Cys-X-Cys), member 10; small inducible cytokine subfamily B (Cys-X-Cys), member 10; small-inducible cytokine B10
Common Name CXCL10 (IP-10)
Gene Symbol CXCL10
Biological Activity Determined by its ability to chemoattract human T-Lymphocytes using a concentration of 10-50 ng/ml
Conjugate Unconjugated
Recombinant Recombinant
Sequence VPLSRTVRCT CISISNQPVN PRSLEKLEII PASQFCPRVE IIATMKKKGE KRCLNPESKA IKNLLKAVSK ERSKRSP
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.