Learn More
Description
Specifications
Specifications
| Assay Range | 156.00 - 10000pg/mL |
| Assay Sensitivity | 47pg/mL |
| Conjugate | HRP |
| Cross Reactivity | Cross-species reactivity not tested. Cross-reactivity observed with 1 or more available related molecules |
| Product Type | Solid Phase Sandwich ELISA |
| Sample Type | Cell Culture Supernatant, Plasma (EDTA), Platelet-poor Plasma (Heparin) |
| Specificity | Natural and Recombinant SDF-1 alpha |
| Sufficient For | 1 x 96-well Plate |
| Target Species | Human |
| For Use With (Application) | ELISA |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
