Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human CXCL12/SDF-1 alpha Quantikine ELISA Kit

Catalog No. SSA00
Change view
Click to view available options
Quantity:
1 Plate
6 Plates
Sufficient For:
1 x 96-well Plate
6 x 96-well Plates
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Sufficient For
SSA00 6 Plates 6 x 96-well Plates
DSA00 1 Plate 1 x 96-well Plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. SSA00 Supplier R&D Systems™ Supplier No. SSA00
Add to Cart
Add to Cart
This item is not returnable. View return policy

Human CXCL12/SDF-1 alpha Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human CXCL12/SDF-1 alpha Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human CXCL12/SDF-1 alpha in cell culture supernates (10 ul), edta plasma (100 ul), platelet-poor heparin plasma (100 ul).

Specifications

Assay Range 156.00 - 10000pg/mL
Assay Sensitivity 47pg/mL
Conjugate HRP
Cross Reactivity Cross-species reactivity not tested. Cross-reactivity observed with 1 or more available related molecules
Product Type Solid Phase Sandwich ELISA
Sample Type Cell Culture Supernatant, Plasma (EDTA), Platelet-poor Plasma (Heparin)
Specificity Natural and Recombinant SDF-1 alpha
Sufficient For 6 x 96-well Plates
Target Species Human
For Use With (Application) ELISA
Gene ID (Entrez) 80381
Quantity 6 Plates
Sample Volume Cell Culture Supernates (10uL),EDTA Plasma (100uL),Platelet-poor Heparin Plasma (100uL)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Target B7-H3/CD276
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.