Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human CXCL2 (GRO beta) Recombinant Protein, PeproTech®

Catalog No. 30039500UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 μg
100 μg
2 μg
250 μg
500 μg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
30039500UG 500 μg
30039100UG 100 μg
300-39-10UG 10 μg
30039250UG 250 μg
300-39-2UG 2 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 30039500UG Supplier Gibco™ Supplier No. 30039500UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

300-39-1MG will be provided as 2 x 500 μg (300-39-500UG). Recombinant Human GRO-beta is a 7.9 kDa protein consisting of 73 amino acids, including the 'ELR' motif common to the CXC chemokine family that binds to CXCR1 or CXCR2. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

Produced by activated monocytes and neutrophils and expressed at sites of inflammation. Hematoregulatory chemokine, which, in vitro, suppresses hematopoietic progenitor cell proliferation. GRO-beta(5-73) shows a highly enhanced hematopoietic activity.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P19875
For Use With (Application) Control, Western Blot Control, ELISA Standard, Bioactivity
Formulation Protein with no preservative
Gene ID (Entrez) 2920
Molecular Weight (g/mol) 7.9 kDa
Name Human CXCL2 (GRO beta)
Purification Method Purified
Quantity 500 μg
Source E. coli
Storage Requirements -20°C
Regulatory Status RUO
Endotoxin Concentration <1 EU/μg
Gene Alias C Cmotif chemokine; C X C motif chemokine; CC motif chemokine; CCmotif chemokine; chemokine (C-X-C motif) ligand 2; CINC-2a; Cinc3; CINC-3; CXC; CXC motif chemokine; C-X-C motif chemokine 2; C-X-C motif chemokine ligand 2; CXCL; CXCL2; Cytokine-induced neutrophil chemoattractant 3; GRO b; gro beta; GRO β; GRO2; GRO2 oncogene; GROb; GRObeta; gro-beta; GRO-beta(5-73); GRO-beta-T; Growth-regulated protein beta; GROβ; Hematopoietic synergistic factor; HSF; Macrophage inflammatory protein 2; macrophage inflammatory protein 2-alpha; melanoma growth stimulatory activity beta; MGSA beta; Mgsa-b; MIP 2 a; MIP 2 alpha; MIP 2 α; Mip2; MIP-2; MIP2 a; MIP2 α; MIP2A; MIP-2a; MIP2-alpha; SB-251353; Scyb; Scyb2; small inducible cytokine subfamily, member 2
Common Name CXCL2
Gene Symbol Cxcl2
Biological Activity Determined by its ability to chemoattract CXCR2 transfected 293 cells using a concentration range of 10.0-100.0 ng/ml
Product Type Protein
Conjugate Unconjugated
Recombinant Recombinant
Sequence APLATELRCQ CLQTLQGIHL KNIQSVKVKS PGPHCAQTEV IATLKNGQKA CLNPASPMVK KIIEKMLKNG KSN
Content And Storage -20°C
Activity Determined by its ability to chemoattract CXCR2 transfected 293 cells using a concentration range of 10.0-100.0 ng/ml.
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.