Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human CXCL5/ENA-78 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB654500
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB654500 500 μg
MAB654SP 25 μg
MAB654100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. MAB654500 Supplier R&D Systems Supplier No. MAB654500
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

CXCL5/ENA-78 Monoclonal specifically detects CXCL5/ENA-78 in Human samples. It is validated for Neutralization, Intracellular Staining by Flow Cytometry, CyTOF-ready, ELISA Capture (Matched Antibody Pair).

Specifications

Antigen CXCL5/ENA-78
Applications Neutralization, Flow Cytometry, CyTOF, ELISA
Classification Monoclonal
Clone 33170
Conjugate Unconjugated
Dilution Neutralization 0.65 to 5 μg/mL, Intracellular Staining by Flow Cytometry 0.25 μg/10^6 cells, CyTOF-ready, ELISA Capture (Matched Antibody Pair) 2 to 8 μg/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P42830
Gene Alias chemokine (C-C motif) ligand 5, C-X-C motif chemokine 5, CXCL5, ENA78, ENA-78, ENA-78(1-78), Epithelial-derived neutrophil-activating protein 78, Neutrophil-activating peptide ENA-78, neutrophil-activating protein 78, SCYB5ENA78
Gene Symbols CXCL5
Host Species Mouse
Immunogen E. coli-derived recombinant human CXCL5/ENA-78 Ala37-Asn114 Accession # P42830
Purification Method Protein A or G purified from ascites
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6374
Test Specificity Detects human CXCL5/ENA-78 in ELISAs. Does not cross-react with recombinant human CXCL1, 2, or 3.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Ascites
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.