Learn More
Kingfisher Biotech Inc Human CXCL7 Recombinant Protein
SDP
Supplier:  Kingfisher Biotech Inc RP1716H100
Encompass_Preferred
The Human CXCL7 yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Human CXCL7 applications are for cell culture, ELISA standard, and Western Blot Control. Human CXCL7 yeast-derived recombinant protein can be purchased in multiple sizes. Human CXCL7 Specifications: (Molecular Weight: 10.6 kDa) (Amino Acid Sequence: TALASSTKGQ TKRNLAKGKE ESLDSDLYAE LRCMCIKTTS GIHPKNIQSL EVIGKGTHCN QVEVIATLKD GRKICLDPDA PRIKKIVQKK LAGDESAD (98)) (Gene ID: 5473). For research use only. Made in the USA
Catalog No. 50-253-2892
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.