Learn More
ACROBiosystems Human CXCR3 500ug
Supplier:  ACROBiosystems CX3H52D5500UG
Encompass_Preferred
Human CXCR3 Protein FlagHis Tag (CX3-H52D5) is expressed from human 293 cells (HEK293). It contains AA Val 2 - Leu 368 (Accession # P49682-1).
Catalog No. 50-253-5190
Only null left
Add to Cart
Request a Quote
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.