Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Cyclin E1 Antibody, R&D Systems™

Catalog No. MAB68101100
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB68101100 100 μg
MAB68101SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB68101100 Supplier R&D Systems Supplier No. MAB68101100
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Cyclin E1 Monoclonal specifically detects Cyclin E1 in Human samples. It is validated for Western Blot.

Specifications

Antigen Cyclin E1
Applications Western Blot
Classification Monoclonal
Clone 713548
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Accession No. P24864
Gene Alias CCNE, CCNE1, CCNEcyclin Es, cyclin E1, cyclin Et, G1/S-specific cyclin-E1
Host Species Mouse
Immunogen E. coli-derived recombinant human Cyclin E1 Met16-Lys143 Accession # P24864
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 898
Test Specificity Detects human Cyclin E1 in direct ELISAs and Western blots.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.