Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human/Cynomolgus Monkey PD-L1/B7-H1 Quantikine ELISA Kit
SDP

Catalog No. DB7H10 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DB7H10 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DB7H10 Supplier R&D Systems™ Supplier No. DB7H10
Save 15% on Immunology Workflow Reagents
Use promo code IMMUNO15
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy

Human PD-L1 Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility

The Human PD-L1 Quantikine ELISA Kit quantifies human, cynomolgus monkey PD-L1 in cell culture supernates (100 μL), cell lysates (100 μL), serum (100 μL), edta plasma (100 μL), heparin plasma (100 μL), urine (100 μL).

Specifications

Assay Range 25.00 to 1600 pg/mL
Assay Sensitivity 4.52 pg/mL
Product Type ELISA
Sample Type Cell Culture Supernatant, Cell Lysates, Plasma (EDTA, Heparin), Serum, Urine
Specificity Natural and Recombinant Human B7-H1/PD-L1. This assay also recognizes natural cynomolgus monkey B7-H1/PD-L1.
For Use With (Application) ELISA
Gene ID (Entrez) 29126
Host Species Human, Cynomolgus Monkey
Quantity 1 Kit
Storage Requirements Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Synonym Avelumab, B7-H, B7H1, B7-H1, B7H1PDCD1L1, CD274, CD274 antigenMGC142294, CD274 molecule, PDCD1L1, PDCD1LG1, PDCD1LG1MGC142296, PDL1, PD-L1, PD-L1B7 homolog 1, PDL1PDCD1 ligand 1, programmed cell death 1 ligand 1, Programmed death ligand 1
Target PD-L1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.