Learn More
Specifications
Specifications
| Gene | CYP21A2 |
| Gene ID (Entrez) | 1589 |
| Gene Alias | 21-OHase, CA21HCYP21B, CAH1, CPS1, CYP21EC 1.14.99.10, Cytochrome P450 21, Cytochrome P450 XXI, cytochrome P450, family 21, subfamily A, polypeptide 2, cytochrome P450, subfamily XXIA (steroid 21-hydroxylase, congenital adrenalhyperplasia), polypeptide 2, Cytochrome P-450c21, Cytochrome P450-C21, Cytochrome P450-C21B, EC 1.14.99, MGC150536, MGC150537, P450c21B, steroid 21-hydroxylase, steroid 21-monooxygenase |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Product Line | R&D Systems |
| Quantity | 10 μg |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
