Learn More
ACROBiosystems ACROBiosystems Human Cystatin SA / CST2 Protein, His Tag
Supplier:  ACROBiosystems CS2H522450UG
Encompass_Preferred
Recombinant Protein;50UG;Human Cystatin SA, His Tag (CS2-H5224) is expressed from human 293 cells (HEK293). It contains AA Trp 21 - Ala 141 (Accession # AAH62679).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 15.2 kDa. The protein migrates as 15-16 kDa under reducing (R) condition (SDS-PAGE).
Catalog No. 50-210-6613
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.