Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human Cytochrome c Quantikine ELISA Kit
GSA_VASDP

Catalog No. SCTC0 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
1 Plate
6 Plates
Sufficient For:
1 x 96-well Plate
6 x 96-well Plates
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Sufficient For
SCTC0 6 Plates 6 x 96-well Plates
DCTC0 1 Plate 1 x 96-well Plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. SCTC0 Supplier R&D Systems™ Supplier No. SCTC0
Add to Cart
Add to Cart
This item is not returnable. View return policy

Human Cytochrome c Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human Cytochrome c Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human Cytochrome c in cell lysates (20 ul), subcellular fractions (100 ul).

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Assay Range 0.63 - 20ng/mL
Assay Sensitivity 0.31ng/mL
Conjugate HRP
Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules
Product Type Solid Phase Sandwich ELISA
Sample Type Cell Lysates, Subcellular Fractions
Specificity Natural Human Cytochrome C
Sufficient For 6 x 96-well Plates
Target Species Human
For Use With (Application) ELISA
Gene ID (Entrez) 54205
Quantity 6 Plates
Sample Volume Cell Lysates (20uL),Subcellular Fractions (100uL)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Synonym CYCHCS, CYCS, cytochrome c, cytochrome c, somatic, THC4
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.