Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human Cytokeratin 19/KRT19 ELISA Kit

Catalog No. EH146RB
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EH146RB 96 Tests
EH146RBX5 5 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. EH146RB Supplier Invitrogen™ Supplier No. EH146RB
Add to Cart
Add to Cart

ELISA

Human CytokeRatin 19/KRT19 quantitates human Cytokeratin 19/KRT19 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human Cytokeratin 19/KRT19.

Cytokeratin 19 is part of a subfamily of intermediate filament proteins and are characterized by a remarkable biochemical diversity, represented in human epithelial tissues by at least 20 different polypeptides. Cytokeratins range in molecular weight from 40-68 kDa and isoelectric pH between 4.9 - 7.8. The individual human cytokeratins are numbered 1 to 20. The various epithelia in the human body usually express cytokeratins which are not only characteristic of the type of epithelium, but also related to the degree of maturation or differentiation within an epithelium. Cytokeratin subtype expression patterns are used to an increasing extent in the distinction of different types of epithelial malignancies. The cytokeratin antibodies are not only of assistance in the differential diagnosis of tumors using immunohistochemistry on tissue sections, but are also a useful tool in cytopathology and flow cytometric assays. For example, cytokeratin 19 is not expressed in hepatocytes, therefore, it is useful in the identification of liver metastasis.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number 40-kDa keratin intermediate filament; AI663979; C K1CS; CK 19; CK19; CK-19; cytokeratin 19; cytokeratin-19; EndoC; K19; K1CS; Ka19; keratin 19; keratin 19, type I; keratin complex 1, acidic, gene 19; keratin, type I cytoskeletal 19; keratin, type I, 40-kd; keratin-19; Krt-1.19; Krt1-19; KRT19; MGC15366; Type I keratin Ka19
Assay Range 0.061 to 15 ng/mL
Assay Sensitivity 0.061 ng/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 3880
Gene Symbol KRT19
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Storage Requirements 2°C to 8°C
Target Cytokeratin 19
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.