Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human Cytokeratin 8/KRT8 ELISA Kit

Catalog No. EH147RBX10
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 x 96 Tests
5 x 96 Tests
96 Tests
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EH147RBX10 10 x 96 Tests
EH147RBX5 5 x 96 Tests
EH147RB 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. EH147RBX10 Supplier Invitrogen™ Supplier No. EH147RBX10
Add to Cart
Add to Cart
Request a Quote

ELISA

Human CytokeRatin 8/KRT8 quantitates human Cytokeratin 8/KRT8 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human Cytokeratin 8/KRT8.

Cytokeratin 8 is a member of the type II keratin family clustered on the long arm of chromosome 12. Type I and type II keratins heteropolymerize to form intermediate-sized filaments in the cytoplasm of epithelial cells. Cutokeratin 8 typically dimerizes with keratin 18 to form an intermediate filament in simple single-layered epithelial cells. This protein plays a role in maintaining cellular structural integrity and also functions in signal transduction and cellular differentiation. Mutations in this gene cause cryptogenic cirrhosis.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number AA960620; AL022697; AU019895; C CY K2C8; CARD2; cb186; CK 8; CK8; CK-8; CYK8; CYKER; cytokeratin 8; cytokeratin 8 (370 AA); Cytokeratin endo A; cytokeratin-8; Cytokeratin-A; DreK8; EndoA; etID33550.23; etID43142.23; etID43142.8; hard keratin; K2C8; K8; K-8; KERA; keratin 8; keratin 8, type II; keratin complex 2, basic, gene 8; keratin, type 2, gene 8; keratin, type II cytoskeletal 8; Keratin-8; KO; Krt-2.8; krt2-8; KRT8; LOW QUALITY PROTEIN: keratin, type II cytoskeletal 8; sb:cb186; type II cytokeratin A (no0.8); type-II keratin Kb8; wu:fa20h05; wu:fa95h10; wu:fb96c06; zfk8; zf-K8
Assay Range 0.410 to 100 pg/mL
Assay Sensitivity 0.41 pg/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 3856
Gene Symbol KRT8
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 10 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Storage Requirements 2°C to 8°C
Target Cytokeratin 8
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.