Learn More
Abcam Human DAP12 (pY91) ELISA Kit, 1 x 96 Tests
Supplier:  Abcam AB3161331X96TESTS
Encompass_Preferred
Human DAP12 (pY91) ELISA Kit is a single-wash 90-min Simplestep used to detect Human DAP12 (pY91). The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Semi-quantitative Sandwich ELISA - 450 nm readout : works on any standard plate reader
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-3345
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.