Learn More
Human DC-SIGN/CD209 Alexa Fluor™ 700-conjugated Antibody, R&D Systems™
Description
Specifications
Specifications
| Antigen | DC-SIGN/CD209 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 120507 |
| Conjugate | Alexa Fluor 700 |
| Dilution | Flow Cytometry 5 uL/10^6 cells |
| Formulation | Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide |
| Gene Alias | CD209, CD209 antigendendritic cell-specific intracellular adhesion molecules (ICAM)-3 grabbingnon-integrin, CD209 molecule, CDSIGNHIV gpl20-binding protein, CLEC4L, CLEC4LC-type lectin domain family 4 member L, DCSIGN, DC-SIGN1, DC-SIGN1C-type lectin domain family 4, member L, DC-SIGNMGC129965, Dendritic cell-specific ICAM-3-grabbing non-integrin 1 |
| Gene Symbols | CD209 |
| Host Species | Mouse |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.