Learn More
Invitrogen™ Human DCBLD2/ESDN ELISA Kit
Description
Human DCBLD2/ESDN quantitates human DCBLD2/ESDN in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human DCBLD2/ESDN.
Specifications
Specifications
| Accession Number | 1700055P21Rik; AW146002; CLCP1; coagulation factor V/VIII-homology domains protein 1; CUB, LCCL and coagulation factor V/VIII-homology domains protein 1; Dcbld2; discoidin; discoidin, CUB and LCCL domain containing 2; discoidin, CUB and LCCL domain-containing protein 2; Endothelial and smooth muscle cell-derived neuropilin-like protein; Esdn |
| Assay Range | 0.041 to 10 ng/mL |
| Assay Sensitivity | 0.041 ng/mL |
| Conjugate | HRP |
| Product Type | ELISA |
| Sample Type | Plasma, Serum, Supernatant |
| For Use With (Equipment) | Colorimetric Microplate Reader |
| Gene ID (Entrez) | 131566 |
| Gene Symbol | DCBLD2 |
| Interassay CV | <12% |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.