Learn More
BioVendor, LLC Human DD/E FDP Plm Generated, Mouse Monoclonal Antibody (100 ug)
SDP
Supplier:  BioVendor, LLC QZBFBDDE2100
Encompass
Human DD/E FDP Plm Generated, Mouse Monoclonal Antibody (100 ug). Manufactured by QuickZyme Biosciences. Mouse MAb against human DD/E complex (specific for plasmin generated fibrin degredation products). Does not react with fibrinogen, fibrin, or non-cross linked fibrin(ogen) degradation products. Can be used in ELISA or agglutination test for such cross-linked degradation products. Synonyms: D-Dimer DD/E2 Fibrin Split Products, FSP. SUMMARY: Fibrin functions in hemostasis as a primary component of blood clots. It is also important in the early stages of wound repair. Fibrinolysis is the enzymatic breakdown of fibrin in blood clots and results in various Fibrin Degradation Products (FDP). High levels of FDP in plasma indicate abnormal fibrinolytic and/or fibrinogenolytic activity. Elevated FDP levels are typical in disseminated intravascular coagulation (DIC), but are not specific for DIC and are also seen numerous other clinical conditions.
Catalog No. 50-206-9098
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.