Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human DDX10 Full-length ORF (NP_004389.2, 1 a.a. - 875 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89941636 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
2 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-941-636 2 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-941-636 Supplier Abnova™ Supplier No. H00001662P01
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

DEAD box proteins, characterized by the conserved motif Asp-Glu-Ala-Asp (DEAD), are putative RNA helicases. They are implicated in a number of cellular processes involving alteration of RNA secondary structure such as translation initiation, nuclear and mitochondrial splicing, and ribosome and spliceosome assembly. Based on their distribution patterns, some members of this family are believed to be involved in embryogenesis, spermatogenesis, and cellular growth and division. This gene encodes a DEAD box protein, and it may be involved in ribosome assembly. Fusion of this gene and the nucleoporin gene, NUP98, by inversion 11 (p15q22) chromosome translocation is found in the patients with de novo or therapy-related myeloid malignancies. [provided by RefSeq]

Sequence: MGKTANSPGSGARPDPVRSFNRWKKKHSHRQNKKKQLRKQLKKPEWQVERESISRLMQNYEKINVNEITRFSDFPLSKKTLKGLQEAQYRLVTEIQKQTIGLALQGKDVLGAAKTGSGKTLAFLVPVLEALYRLQWTSTDGLGVLIISPTRELAYQTFEVLRKVGKNHDFSAGLIIGGKDLKHEAERINNINILVCTPGRLLQHMDETVSFHATDLQMLVLDEADRILDMGFADTMNAVIENLPKKRQTLLFSATQTKSVKDLARLSLKNPEYVWVHEKAKYSTPATLEQNYIVCELQQKISVLYSFLRSHLKKKSIVFFSSCKEVQYLYRVFCRLRPGVSILALHGRQQQMRRMEVYNEFVRKRAAVLFATDIAARGLDFPAVNWVLQFDCPEDANTYIHRAGRTARYKEDGEALLILLPSEKAMVQQLLQKKVPVKEIKINPEKLIDVQKKLESILAQDQDLKERAQRCFVSYVRSVYLMKDKEVFDVSKLPIPEYALSLGLAVAPRVRFLQKMQKQPTKELVRSQADKVIEPRAPSLTNDEVEEFRAYFNEKMSILQKGGKRLEGTEHRQDNDTGNEEQEEEEDDEEEMEEKLAKAKGSQAPSLPNTSEAQKIKEVPTQFLDRDEEEEDADFLKVKRHNVFGLDLKDEKTLQKKEPSKSSIKKKMTKVAEAKKVMKRNFKVNKKITFTDEGELVQQWPQMQKSAIKDAEEDDDTGGINLHKAKERLQEEDKFDKEEYRKKIKAKHREKRLKEREARREANKRQAKAKDEEEAFLDWSDDDDDDDDGFDPSTLPDPDKYRSSEDSDSEDMENKISDTKKKQGMKKRSNSEVEDVGPTSHNRKKARWDTLEPLDTGLSLAEDEELVLHLLRSQS

Specifications

Accession Number NP_004389.2
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 1662
Molecular Weight (g/mol) 127.3kDa
Name DDX10 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 2 μg
Immunogen MGKTANSPGSGARPDPVRSFNRWKKKHSHRQNKKKQLRKQLKKPEWQVERESISRLMQNYEKINVNEITRFSDFPLSKKTLKGLQEAQYRLVTEIQKQTIGLALQGKDVLGAAKTGSGKTLAFLVPVLEALYRLQWTSTDGLGVLIISPTRELAYQTFEVLRKVGKNHDFSAGLIIGGKDLKHEAERINNINILVCTPGRLLQHMDETVSFHATDLQMLVLDEADRILDMGFADTMNAVIENLPKKRQTLLFSATQTKSVKDLARLSLKNPEYVWVHEKAKYSTPATLEQNYIVCELQQKISVLYSFLRSHLKKKSIVFFSSCKEVQYLYRVFCRLRPGVSILALHGRQQQMRRMEVYNEFVRKRAAVLFATDIAARGLDFPAVNWVLQFDCPEDANTYIHRAGRTARYKEDGEALLILLPSEKAMVQQLLQKKVPVKEIKINPEKLIDVQKKLESILAQDQDLKERAQRCFVSYVRSVYLMKDKEVFDVSKLPIPEYALSLGLAVAPRVRFLQKMQKQPTKELVRSQADKVIEPRAPSLTNDEVEEFRAYFNEKMSILQKGGKRLEGTEHRQDNDTGNEEQEEEEDDEEEMEEKLAKAKGSQAPSLPNTSEAQKIKEVPTQFLDRDEEEEDADFLKVKRHNVFGLDLKDEKTLQKKEPSKSSIKKKMTKVAEAKKVMKRNFKVNKKITFTDEGELVQQWPQMQKSAIKDAEEDDDTGGINLHKAKERLQEEDKFDKEEYRKKIKAKHREKRLKEREARREANKRQAKAKDEEEAFLDWSDDDDDDDDGFDPSTLPDPDKYRSSEDSDSEDMENKISDTKKKQGMKKRSNSEVEDVGPTSHNRKKARWDTLEPLDTGLSLAEDEELVLHLLRSQS
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias HRH-J8
Common Name DDX10
Gene Symbol DDX10
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.