Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human DDX41 Synthetic Peptide

Catalog No. PEP1593
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PEP1593 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PEP1593 Supplier Invitrogen™ Supplier No. PEP1593
Add to Cart
Add to Cart

Synthetic Peptide

DDX41 Synthetic Peptide, PEP-1593, from Invitrogen.

Probable ATP-dependent RNA helicase DDX41 is a DEAD box protein characterized by the conserved motif Asp-Glu-Ala-Asp. It is required during post-transcriptional gene expression and may be involved in pre-mRNA splicing. Mutations affecting the gene can result in myeloproliferative/lymphoproliferative neoplasms, familial (MPLPF).
TRUSTED_SUSTAINABILITY

Specifications

Gene ID (Entrez) 51428
Species Human
Purification Method purified
Accession Number Q9UJV9
Common Name Human DDX41
Concentration 200 μg/mL
Content And Storage -20°C
Description Synthetic Peptide
Format Liquid
Formulation PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2
For Use With (Application) Blocking Assay, Control
Gene Symbol DDX41
Lyophilized No
Preservative 0.02% sodium azide
Product Type Peptide
Quantity 50 μg
Regulatory Status RUO - research use only
Storage Buffer PBS
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.