Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human Defensin alpha 5 ELISA Kit (Chemiluminescence)
SDP

Catalog No. NB295273 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB295273 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB295273 Supplier R&D Systems™ Supplier No. NBP324603

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems.

Specifications

Assay Range 7.81 to 500 pg/mL
Assay Sensitivity 4.69 pg/mL
Product Line Novus
Product Type ELISA Kit (Chemiluminescence)
Sample Type Serum, Plasma, Other Biological Fluids
Specificity This kit recognizes Human Defensin Alpha 5 in samples. No significant cross-reactivity or interference between Human Defensin Alpha 5 and analogues was observed.
For Use With (Application) ELISA, Sandwich ELISA
Gene ID (Entrez) 1670
Gene Symbol DEFA5
Quantity 1 Kit
Research Discipline Immunology, Innate Immunity
Storage Requirements Storage of components varies. See protocol for specific instructions.
Synonym DEF5defensin, alpha 5, preproprotein, defensin 5, Defensin, alpha 5, defensin, alpha 5, Paneth cell-specific, defensin-5, HD-5, MGC129728
Target Human
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.