Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human DEP-1/CD148 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB19341SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB19341SP 25 μg
MAB19341 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB19341SP Supplier R&D Systems Supplier No. MAB19341SP
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

DEP-1/CD148 Monoclonal specifically detects DEP-1/CD148 in Human samples. It is validated for Flow Cytometry, CyTOF-ready.

Specifications

Antigen DEP-1/CD148
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 261922
Concentration LYOPH
Conjugate Unconjugated
Dilution Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q12913
Gene Alias CD148, CD148 antigen, Density-enhanced phosphatase 1, DEP1, DEP-1, DEP1EC 3.1.3.48, HPTP eta, HPTPeta, HPTP-eta, human density enhanced phosphatase-1, protein tyrosine phosphatase, receptor type, J, protein tyrosine phosphatase, receptor type, J polypeptide, Protein-tyrosine phosphatase eta, Protein-tyrosine phosphatase receptor type J, PTPRJ, receptor-type tyrosine-protein phosphatase eta, R-PTP-eta, R-PTP-J, SCC1, susceptibility to colon cancer 1, mouse, homolog of
Gene Symbols PTPRJ
Host Species Mouse
Immunogen E. coli-derived recombinant human DEP-1/CD148 Arg997-Ala1337 Accession # Q12913
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5795
Test Specificity Detects human DEP-1/CD148.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.