Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human DFFB Partial ORF (NP_004393, 229 a.a. - 338 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89941827 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 ug
25 ug
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-941-827 10 ug
89-941-828 25 ug
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-941-827 Supplier Abnova™ Supplier No. H00001677Q01S
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

Apoptosis is a cell death process that removes toxic and/or useless cells during mammalian development. The apoptotic process is accompanied by shrinkage and fragmentation of the cells and nuclei and degradation of the chromosomal DNA into nucleosomal units. DNA fragmentation factor (DFF) is a heterodimeric protein of 40-kD (DFFB) and 45-kD (DFFA) subunits. DFFA is the substrate for caspase-3 and triggers DNA fragmentation during apoptosis. DFF becomes activated when DFFA is cleaved by caspase-3. The cleaved fragments of DFFA dissociate from DFFB, the active component of DFF. DFFB has been found to trigger both DNA fragmentation and chromatin condensation during apoptosis. Alternatively spliced transcript variants encoding distinct isoforms have been found for this gene but the biological validity of these variants has not been determined. [provided by RefSeq]

Sequence: PFDMDSCLSRHSINPYSNRESRILFSTWNLDHIIEKKRTIIPTLVEAIKEQDGREVDWEYFYGLLFTSENLKLVHIVCHKKTTHKLNCDPSRIYKPQTRLKRKQPVRKRQ

Specifications

Accession Number NP_004393
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 1677
Molecular Weight (g/mol) 37.84kDa
Name DFFB (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 ug
Immunogen PFDMDSCLSRHSINPYSNRESRILFSTWNLDHIIEKKRTIIPTLVEAIKEQDGREVDWEYFYGLLFTSENLKLVHIVCHKKTTHKLNCDPSRIYKPQTRLKRKQPVRKRQ
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias CAD/CPAN/DFF-40/DFF2/DFF40
Common Name DFFB
Gene Symbol DFFB
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.