Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human DGAT1 (NP_036211) VersaClone cDNA
SDP

Catalog No. RDC2928 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RDC2928 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. RDC2928 Supplier R&D Systems™ Supplier No. RDC2928
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Contains gene inserts that are fully sequenced and produced with high quality

Specifications

Gene DGAT1
Accession Number NP_036211
Gene ID (Entrez) 8694
Gene Alias ACAT-related gene product 1, AGRP1, ARAT, ARGP1, ARGP1acyl coenzyme A:cholesterol acyltransferase related gene 1, DGAT, DGATACAT related gene product 1, diacylglycerol O-acyltransferase 1, diacylglycerol O-acyltransferase homolog 1, diacylglycerol O-acyltransferase homolog 1 (mouse), DIAR7, Diglyceride acyltransferase, EC 2.3.1, EC 2.3.1.20
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Species Human
Product Line R&D Systems
Quantity 10 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.