Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human DGCR6 Full-length ORF (NP_005666.2, 1 a.a. - 220 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89941850 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-941-850 10 μg
89-941-851 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-941-850 Supplier Abnova™ Supplier No. H00008214P01S
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

DiGeorge syndrome, and more widely, the CATCH 22 syndrome, are associated with microdeletions in chromosomal region 22q11.2. The product of this gene shares homology with the Drosophila melanogaster gonadal protein, which participates in gonadal and germ cell development, and with the gamma-1 subunit of human laminin. This gene is a candidate for involvement in DiGeorge syndrome pathology and in schizophrenia. [provided by RefSeq]

Sequence: MERYAGALEEVADGARQQERHYQLLSALQSLVKELPSSFQQRLSYTTLSDLALALLDGTVFEIVQGLLEIQHLTEKSLYNQRLRLQNEHRVLRQALRQKHQEAQQACRPHNLPVLQAAQQRELEAVEHRIREEQRAMDQKIVLELDRKVADQQSTLEKAGVAGFYVTTNPQELMLQMNLLELIRKLQQRGCWAGKAALGLGGPWQLPAAQCDQKGSPVPP

Specifications

Accession Number NP_005666.2
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 8214
Molecular Weight (g/mol) 51.4kDa
Name DGCR6 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 μg
Immunogen MERYAGALEEVADGARQQERHYQLLSALQSLVKELPSSFQQRLSYTTLSDLALALLDGTVFEIVQGLLEIQHLTEKSLYNQRLRLQNEHRVLRQALRQKHQEAQQACRPHNLPVLQAAQQRELEAVEHRIREEQRAMDQKIVLELDRKVADQQSTLEKAGVAGFYVTTNPQELMLQMNLLELIRKLQQRGCWAGKAALGLGGPWQLPAAQCDQKGSPVPP
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Common Name DGCR6
Gene Symbol DGCR6
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.