Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human DIABLO (NP_063940.1, 1 a.a. - 239 a.a.) Full-length Recombinant Protein with His-GST-tag

Catalog No. 89941966 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-941-966 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-941-966 Supplier Abnova™ Supplier No. P5184
Add to Cart
Add to Cart

Used for ELISA, SDS-PAGE, WB

This gene encodes an inhibitor of apoptosis protein (IAP)-binding protein. The encoded mitochondrial protein enters the cytosol when cells undergo apoptosis, and it moderates the caspase inhibition of IAPs. Multiple polyadenylation sites have been found for this gene. Four alternatively spliced transcript variants have been described for this gene, with two of them encoding different isoforms and the other two probably not encoding a protein. [provided by RefSeq]

Specifications

Accession Number NP_063940.1
For Use With (Application) ELISA, SDS-PAGE, Western Blot
Formulation Liquid
Gene ID (Entrez) 56616
Molecular Weight (g/mol) 56.1kDa
Name DIABLO (Human) Recombinant Protein
Preparation Method Escherichia coli expression system
Quality Control Testing 10% SDS-PAGE Stained with Coomassie Blue
Quantity 50 μg
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias DIABLO-S/FLJ10537/FLJ25049/SMAC/SMAC3
Common Name DIABLO
Gene Symbol DIABLO
Species E. coli
Recombinant Recombinant
Protein Tag His-GST
Expression System Escherichia coli expression system
Form Liquid
Purity or Quality Grade >90%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.