Learn More
Human Dihydrolipoamide Dehydrogenase/DLD Antibody, R&D Systems™

Description
Specifications
Specifications
| Antigen | Dihydrolipoamide Dehydrogenase/DLD |
| Applications | Immunohistochemistry, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 1016619 |
| Conjugate | Unconjugated |
| Dilution | Immunohistochemistry 5-25 ug/mL, Immunocytochemistry 5-25 ug/mL |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. |
| Gene Accession No. | P09622 |
| Gene Alias | 2-oxo-glutarate complex, branched chain keto acid dehydrogenase complex), branched chain keto acid dehydrogenase complex, Diaphorase, dihydrolipoamide dehydrogenase (E3 component of pyruvate dehydrogenase complex, dihydrolipoamide dehydrogenasemitochondrial, DLD, DLDD, DLDH, E3, E3 component of pyruvate dehydrogenase complex, 2-oxo-glutarate complex, EC 1.8.1, EC 1.8.1.4, GCSL, Glycine cleavage system L protein, glycine cleavage system protein L, LAD, Lipoamide Dehydrogenase, lipoamide reductase, lipoyl dehydrogenase, PHE3 |
| Host Species | Mouse |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.