Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human DISP2 Synthetic Peptide

Catalog No. PEP1411
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PEP1411 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PEP1411 Supplier Invitrogen™ Supplier No. PEP1411
Add to Cart
Add to Cart

Synthetic Peptide

DISP2 Synthetic Peptide, PEP-1411, from Invitrogen.

DISP2, also known as dispatched homolog 2, is a protein that plays a critical role in the regulation of the hedgehog signaling pathway. It is involved in the transport and release of hedgehog ligands, which are important for cellular differentiation and tissue development. Mutations in the DISP2 gene have been associated with various developmental disorders, including holoprosencephaly and cleft lip and palate. DISP2 is also being studied as a potential therapeutic target in cancer, as the hedgehog pathway is frequently dysregulated in various types of tumors
TRUSTED_SUSTAINABILITY

Specifications

Gene ID (Entrez) 85455
Species Human
Purification Method purified
Accession Number A7MBM2
Common Name Human DISP2
Concentration 200 μg/mL
Content And Storage -20°C
Description Synthetic Peptide
Format Liquid
Formulation PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2
For Use With (Application) Blocking Assay, Control
Gene Symbol DISP2
Lyophilized No
Preservative 0.02% sodium azide
Product Type Peptide
Quantity 50 μg
Regulatory Status RUO - research use only
Storage Buffer PBS
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.