Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human DKFZP781I1119 Full-length ORF (AAH41348.2, 1 a.a. - 522 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89952501 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-952-501 10 μg
89-952-502 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-952-501 Supplier Abnova™ Supplier No. H00166968P01S
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

Sequence: MLVHDYDDERTLEEEEMMDEGKNFSSEIEDLEKEGTMPLEDLLAFYGYEPTIPAVANSSANSSPSELADELPDMTLDKEEIAKDLLSGDDEETQSSADGLTPSVTSHETSDFFPRPLRSNTACDGDKESEVEDVETDSGNSPEDLRKEIMIGLQYQAEIPPYLGEYDGNEKVYENEDQLLWCPDVVLESKVKEYLVETSLRTGSEKIMDRISAGTHTRDNEQALYELLKCNHNIKEAIERYCCNGKASQGMTAWTEEECRSFEHALMLFGKDFHLIQKNKVRTRTVAECVAFYYMWKKSERYDYFAQQTRFGKKRYNHHPGVTDYMDRLVDETEALGGTVNASALTSNRPEPIPDQQLNILNSFTASDLTALTNSVATVCDPTDVNCLDDSFPPLGNTPRGQVNHVPVVTEELLTLPSNGESDCFNLFETGFYHSELNPMNMCSEESERPAKRLKMGIAVPESFMNEVSVNNLGVDFENHTHHITSAKMAVSVADFGSLSANETNGFISAHALHQHAALHSE

Specifications

Accession Number AAH41348.2
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 166968
Molecular Weight (g/mol) 84.9kDa
Name DKFZP781I1119 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 μg
Immunogen MLVHDYDDERTLEEEEMMDEGKNFSSEIEDLEKEGTMPLEDLLAFYGYEPTIPAVANSSANSSPSELADELPDMTLDKEEIAKDLLSGDDEETQSSADGLTPSVTSHETSDFFPRPLRSNTACDGDKESEVEDVETDSGNSPEDLRKEIMIGLQYQAEIPPYLGEYDGNEKVYENEDQLLWCPDVVLESKVKEYLVETSLRTGSEKIMDRISAGTHTRDNEQALYELLKCNHNIKEAIERYCCNGKASQGMTAWTEEECRSFEHALMLFGKDFHLIQKNKVRTRTVAECVAFYYMWKKSERYDYFAQQTRFGKKRYNHHPGVTDYMDRLVDETEALGGTVNASALTSNRPEPIPDQQLNILNSFTASDLTALTNSVATVCDPTDVNCLDDSFPPLGNTPRGQVNHVPVVTEELLTLPSNGESDCFNLFETGFYHSELNPMNMCSEESERPAKRLKMGIAVPESFMNEVSVNNLGVDFENHTHHITSAKMAVSVADFGSLSANETNGFISAHALHQHAALHSE
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias DKFZp686L09111/DKFZp781G1119/DKFZp781I1119/FLJ35954
Common Name MIER3
Gene Symbol MIER3
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.