Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human DKK-2 Recombinant Protein, PeproTech®

Catalog No. 1204550UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
2 μg
250 μg
50 μg
500 μg
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
120-45-50UG 50 μg
12045100UG 100 μg
120-45-10UG 10 μg
120-45-1MG 1 mg
12045250UG 250 μg
120-45-2UG 2 μg
12045500UG 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Catalog No. 120-45-50UG Supplier Gibco™ Supplier No. 1204550UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Recombinant Human DKK-2 expressed in CHO cells is a glycoprotein that has a calculated molecular weight of 25.8 kDa and contains 234 amino acid residues. Due to glycosylation, human DKK-2 migrates at an apparent molecular weight of approximately 31-36 kDa by SDS-PAGE analysis under non-reducing conditions. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

This gene encodes a protein that is a member of the dickkopf family. The secreted protein contains two cysteine rich regions and is involved in embryonic development through its interactions with the Wnt signaling pathway. It can act as either an agonist or antagonist of Wnt/beta-catenin signaling, depending on the cellular context and the presence of the co-factor kremen 2. Activity of this protein is also modulated by binding to the Wnt co-receptor LDL-receptor related protein 6.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9UBU2
For Use With (Application) Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 27123
Molecular Weight (g/mol) 25.8 kDa
Name Human DKK-2
Quantity 50 μg
Source CHO cells
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias dickkopf 2; dickkopf 2 homolog; Dickkopf gene 2; dickkopf homolog 1; dickkopf homolog 2; dickkopf homolog 2 (Xenopus laevis); dickkopf homolog 4; dickkopf related protein-2; dickkopf WNT signaling pathway inhibitor 2; dickkopf-2; Dickkopf-related protein 2; DKK2; DKK-2; Dkk4; hDkk-2; mDkk-2; UNQ682/PRO1316
Common Name DKK2
Gene Symbol DKK2
Biological Activity Determined by its ability to inhibit alkaline phosphatase activity in differentiating MC3T3 E1 cells. The expected ED50 for this effect is 0.5-1.0 ug/ml
Conjugate Unconjugated
Recombinant Recombinant
Sequence SQIGSSRAKL NSIKSSLGGE TPGQAANRSA GMYQGLAFGG SKKGKNLGQA YPCSSDKECE VGRYCHSPHQ GSSACMVCRR KKKRCHRDGM CCPSTRCNNG ICIPVTESIL TPHIPALDGT RHRDRNHGHY SNHDLGWQNL GRPHTKMSHI KGHEGDPCLR SSDCIEGFCC ARHFWTKICK PVLHQGEVCT KQRKKGSHGL EIFQRCDCAK GLSCKVWKDA TYSSKARLHV CQKI
Content And Storage -20°C
Expression System CHO cells
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.