Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human DKK-3 Recombinant Protein, PeproTech®

Catalog No. 120461MG
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
2 μg
250 μg
500 μg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
120-46-1MG 1 mg
12046100UG 100 μg
120-46-10UG 10 μg
12046250UG 250 μg
120-46-2UG 2 μg
12046500UG 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 120-46-1MG Supplier Gibco™ Supplier No. 120461MG
Add to Cart
Add to Cart
Request a Quote

Recombinant Protein

Recombinant Human DKK-3 expressed in CHO cells is a glycoprotein that has a calculated molecular weight of 36.3 kDa and contains 329 amino acid residues. Due to glycosylation, Human DKK-3 migrates at an apparent molecular weight of approximately 39-49 kDa by SDS-PAGE analysis under non-reducing conditions. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

DKK3, like DKK1, DKK2, and DKK4, possesses an N-terminal signal peptide and 2 conserved cysteine-rich domains, which are separated by a linker region and contain 10 cysteine residues each. The second cysteine region has a putative lipid-binding function that may facilitate WNT/DKK interactions at the plasma membrane. The linker region contains 50 to 55 amino acids in DKK1, DKK2, and DKK4, whereas in DKK3 it contains only 12 amino acids. All DKKs have several potential sites for cleavage by furin-type proteases. Northern blot analysis revealed wide expression of the DKK3 transcript, with highest expression in heart, brain, and spinal cord. In situ hybridization reveals highest expression in mouse brain, eye, and heart.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9UBP4
For Use With (Application) Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 27122
Molecular Weight (g/mol) 36.3 kDa
Name Human DKK-3
Quantity 1 mg
Source CHO cells
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias AW061014; C87148; Dickkopf; dickkopf 3; dickkopf 3 homolog; Dickkopf gene 3; dickkopf homolog 3; dickkopf homolog 3 (Xenopus laevis); dickkopf WNT signaling pathway inhibitor 3; Dickkopf3; dickkopf-3; Dickkopf-related protein 3; DKK 3; Dkk3; dkk-3; hDkk 3; hDkk-3; hhDkk3; mDkk-3; p29; regulated in glioma; Reic; RIG; RIG-like 5-6; RIG-like 7-1; UNQ258/PRO295; Xenopμs
Common Name DKK3
Gene Symbol DKK3
Biological Activity Determined by its ability to inhibit alkaline phosphatase activity in differentiating MC3T3 E1 cells. The expected ED50 for this effect is 2.0-4.0 ng/ml
Conjugate Unconjugated
Recombinant Recombinant
Sequence APAPTATSAP VKPGPALSYP QEEATLNEMF REVEELMEDT QHKLRSAVEE MEAEEAAAKA SSEVNLANLP PSYHNETNTD TKVGNNTIHV HREIHKITNN QTGQMVFSET VITSVGDEEG RRSHECIIDE DCGPSMYCQF ASFQYTCQPC RGQRMLCTRD SECCGDQLCV WGHCTKMATR GSNGTICDNQ RDCQPGLCCA FQRGLLFPVC TPLPVEGELC HDPASRLLDL ITWELEPDGA LDRCPCASGL LCQPHSHSLV YVCKPTFVGS RDQDGEILLP REVPDEYEVG SFMEEVRQEL EDLERSLTEE MALREPAAAA AALLGGEEI
Content And Storage -20°C
Expression System CHO cells
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.