Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human DLL-1 (soluble) Recombinant Protein, PeproTech®

Catalog No. 140085UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
100 μg
25 μg
250 μg
5 μg
500 μg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
140-08-5UG 5 μg
14008100UG 100 μg
140-08-1MG 1 mg
14008250UG 250 μg
140-08-25UG 25 μg
14008500UG 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 140-08-5UG Supplier Gibco™ Supplier No. 140085UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

The human DLL-1 gene consists of a 528 amino acid extracellular domain with one DSL domain, eight EGF-like repeats, a 23 amino acid transmembrane domain, and a 155 amino acid cytoplasmic domain. The calculated molecular weight of Recombinant Human sDLL-1 is 56.3 kDa. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

Delta-like protein 1 (DLL1) is a transmembrane protein characterized by epidermal growth factor (EGF)-like repeats. It is a ligand for Notch receptor. It blocks the differentiation of progenitor cells into the B-cell lineage while promoting the emergence of a population of cells with the characteristics of a T-cell/NK-cell precursor.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O00548
For Use With (Application) Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 28514
Molecular Weight (g/mol) 56.3 kDa
Name Human DLL-1 (soluble)
Quantity 5 μg
Source HEK293 cells
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias C-Delta-1; Delta; delta like canonical Notch ligand 1; delta like-1; DELTA1; delta-like 1; delta-like 1 (Drosophila); delta-like protein 1; DL1; Dll1; Dll1-derived cell-associated form; Dll1-EC; Dll1-IC; Dll1-intracellular form; Dll1-soluble form; Dll1-TMIC; Drosophila Delta homolog 1; H-Delta-1; LOW QUALITY PROTEIN: delta-like protein 1; OTTHUMP00000017690; UNQ146/PRO172
Common Name DLL1
Gene Symbol DLL1
Biological Activity Measured by the ability of immobilized sDLL-1 to enhance BMP-2 induced Alkaline Phosphatase activity in C2C12 mouse myoblast cells.
Conjugate Unconjugated
Recombinant Recombinant
Sequence SGVFELKLQE FVNKKGLLGN RNCCRGGAGP PPCACRTFFR VCLKHYQASV SPEPPCTYGS AVTPVLGVDS FSLPDGGGAD SAFSNPIRFP FGFTWPGTFS LIIEALHTDS PDDLATENPE RLISRLATQR HLTVGEEWSQ DLHSSGRTDL KYSYRFVCDE HYYGEGCSVF CRPRDDAFGH FTCGERGEKV CNPGWKGPYC TEPICLPGCD EQHGFCDKPG ECKCRVGWQG RYCDECIRYP GCLHGTCQQP WQCNCQEGWG GLFCNQDLNY CTHHKPCKNG ATCTNTGQGS YTCSCRPGYT GATCELGIDE CDPSPCKNGG SCTDLENSYS CTCPPGFYGK ICELSAMTCA DGPCFNGGRC SDSPDGGYSC RCPVGYSGFN CEKKIDYCSS SPCSNGAKCV DLGDAYLCRC QAGFSGRHCD DNVDDCASSP CANGGTCRDG VNDFSCTCPP GYTGRNCSAP VSRCEHAPCH NGATCHERGH RYVCECARGY GGPNCQFLLP ELPPGPAVVD LTEKLEGQGG PF
Content And Storage -20°C
Expression System HEK293 cells
Form Lyophilized
Purity or Quality Grade ≥ 95% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.