Learn More
ACROBiosystems resDetect™ Biotinylated Human DLL4 ELISA Kit (Residue Testing)
Supplier:  ACROBiosystems RESA02296TESTS
Encompass_Preferred
Biotinylated Human DLL4 ELISA Kit is based on ELISA sandwich method and designed to measure biotinylated human DLL4 levels in cell culture supernates, serum, and plasma. It contains recombinant biotinylated human DLL4 and a pair of antibodies against the recombinant factor, which are provided by ACROBiosystems. Results are obtained by four parameter logistic curve that were parallel to the standard curves obtained. The verification results indicate that this kit can be used for the quantitative determination of GMP biotinylated human DLL4 (ACROBiosystems, cat#GMP-DL4H23) concentrations. The specificity has been verified.
Catalog No. 50-271-8981
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.