Learn More
CHIMERX Human DNA Polymerase Beta, 200u
Supplier:  CHIMERX 1077 200U
Encompass
Exclusively from CHIMERx, Human DNA Polymerase Beta is a repair enzyme used to fill gaps and nicks in double-stranded DNA.
Catalog No. NC2751907