Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human DNAJB13 Partial ORF (NP_705842, 71 a.a. - 180 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89965900 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-965-900 10 μg
89-965-901 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-965-900 Supplier Abnova™ Supplier No. H00374407Q01S

Used for AP, Array, ELISA, WB-Re

DNAJA13 belongs to the evolutionarily conserved DNAJ/HSP40 family of proteins. For background information on the DNAJ family, see 608375.[supplied by OMIM]

Sequence: GLKGGIPLEFGSQTPWTTGYVFHGKPEKVFHEFFGGNNPFSEFFDAEGSEVDLNFGGLQGRGVKKQDPQVERDLYLSLEDLFFGCTKKIKISRRVLNEDGYSSTIKDKIL

Specifications

Accession Number NP_705842
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 374407
Molecular Weight (g/mol) 37.62kDa
Name DNAJB13 (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 μg
Immunogen GLKGGIPLEFGSQTPWTTGYVFHGKPEKVFHEFFGGNNPFSEFFDAEGSEVDLNFGGLQGRGVKKQDPQVERDLYLSLEDLFFGCTKKIKISRRVLNEDGYSSTIKDKIL
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias FLJ46748/TSARG5/TSARG6
Common Name DNAJB13
Gene Symbol DNAJB13
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.