Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human DOC2B Full-length ORF (AAI53025.1, 1 a.a. - 412 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89942294 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-942-294 10 μg
89-942-295 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-942-294 Supplier Abnova™ Supplier No. H00008447P01S
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

There are at least two protein isoforms of the Double C2 protein, namely alpha (DOC2A) and beta (DOC2B), which contain two C2-like domains. DOC2A and DOC2B are encoded by different genes; these genes are at times confused with the unrelated DAB2 gene which was initially named DOC-2. DOC2B is expressed ubiquitously and is suggested to be involved in Ca(2+)-dependent intracellular vesicle trafficking in various types of cells. [provided by RefSeq]

Sequence: MTLRRRGEKATISIQEHMAIDVCPGPIRPIKQISDYFPRFPRGLPPDAGPRAAAPPDAPARPAVAGAGRRSPSDGAREDDEDVDQLFGAYGSSPGPSPGPSPARPPAKPPEDEPDADGYESDDCTALGTLDFSLLYDQENNALHCTITKAKGLKPMDHNGLADPYVKLHLLPGASKANKLRTKTLRNTLNPTWNETLTYYGITDEDMIRKTLRISVCDEDKFRHNEFIGETRVPLKKLKPNHTKTFNICLEKQLPVDKTEDKSLEERGRILISLKYSSQKQGLLVGIVRCAHLAAMDANGYSDPYVKTYLRPDVDKKSKHKTAVKKKTLNPEFNEEFCYEIKHGDLAKKSLEVTVWDYDIGKSNDFIGGVVLGIHAKGERLKHWFDCLKNKDKRIERWHTLTSELPGAVLSD

Specifications

Accession Number AAI53025.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 8447
Molecular Weight (g/mol) 72.27kDa
Name DOC2B (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 μg
Immunogen MTLRRRGEKATISIQEHMAIDVCPGPIRPIKQISDYFPRFPRGLPPDAGPRAAAPPDAPARPAVAGAGRRSPSDGAREDDEDVDQLFGAYGSSPGPSPGPSPARPPAKPPEDEPDADGYESDDCTALGTLDFSLLYDQENNALHCTITKAKGLKPMDHNGLADPYVKLHLLPGASKANKLRTKTLRNTLNPTWNETLTYYGITDEDMIRKTLRISVCDEDKFRHNEFIGETRVPLKKLKPNHTKTFNICLEKQLPVDKTEDKSLEERGRILISLKYSSQKQGLLVGIVRCAHLAAMDANGYSDPYVKTYLRPDVDKKSKHKTAVKKKTLNPEFNEEFCYEIKHGDLAKKSLEVTVWDYDIGKSNDFIGGVVLGIHAKGERLKHWFDCLKNKDKRIERWHTLTSELPGAVLSD
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Common Name DOC2B
Gene Symbol DOC2B
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.