Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human DRAK1 Synthetic Peptide

Catalog No. PEP0051
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PEP0051 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PEP0051 Supplier Invitrogen™ Supplier No. PEP0051
Add to Cart
Add to Cart

Synthetic Peptide

DRAK1 Synthetic Peptide, PEP-0051, from Invitrogen.

STK17A (DRAK1) is a serine/threonine kinase localized in the nucleus and involved in apoptosis. Diseases associated with STK17A include Colon Squamous Cell Carcinoma. Among its related pathways are Sweet Taste Signaling. Gene Ontology (GO) annotations related to this gene include transferase activity, transferring phosphorus-containing groups and protein tyrosine kinase activity. An important paralog of this gene is STK17B.
TRUSTED_SUSTAINABILITY

Specifications

Gene ID (Entrez) 9263
Species Human
Purification Method purified
Accession Number Q9UEE5
Common Name Human DRAK1
Concentration 200 μg/mL
Content And Storage -20°C
Description Synthetic Peptide
Format Liquid
Formulation PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2
For Use With (Application) Blocking Assay, Control
Gene Symbol STK17A
Lyophilized No
Preservative 0.02% sodium azide
Product Type Peptide
Quantity 50 μg
Regulatory Status RUO - research use only
Storage Buffer PBS
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.