Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human DUSP9 Full-length ORF (AAH60837.1, 1 a.a. - 384 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89942582 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 ug
25 ug
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-942-582 25 ug
89-942-581 10 ug
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-942-582 Supplier Abnova™ Supplier No. H00001852P01L
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

The protein encoded by this gene is a member of the dual specificity protein phosphatase subfamily. These phosphatases inactivate their target kinases by dephosphorylating both the phosphoserine/threonine and phosphotyrosine residues. They negatively regulate members of the mitogen-activated protein (MAP) kinase superfamily (MAPK/ERK, SAPK/JNK, p38), which is associated with cellular proliferation and differentiation. Different members of the family of dual specificity phosphatases show distinct substrate specificities for various MAP kinases, different tissue distribution and subcellular localization, and different modes of inducibility of their expression by extracellular stimuli. This gene product shows selectivity for members of the ERK family of MAP kinases, is expressed only in placenta, kidney, and fetal liver, and is localized to the cytoplasm and nucleus. [provided by RefSeq]

Sequence: MEGLGRSCLWLRRELSPPRPRLLLLDCRSRELYESARIGGALSVALPALLLRRLRRGSLSVRALLPGPPLQPPPPAPVLLYDQGGGRRRRGEAEAEAEEWEAESVLGTLLQKLREEGYLAYYLQGGFSRFQAECPHLCETSLAGRAGSSMAPLPGPVPVVGLGSLCLGSDCSDAESEADRDSMSCGLDSEGATPPPVGLRASFPVQILPNLYLGSARDSANLESLAKLGIRYILNVTPNLPNFFEKNGDFHYKQIPISDHWSQNLSRFFPEAIEFIDEALSQNRGVLVHCLAGVSRSVTVTVAYLMQKLHLSLNDAYDLVKRKKSNISPNFNFMGQLLDFERSLRLEERHSQEQGSGGQASAASNPPSFFTTPTSDGAFELAPT

Specifications

Accession Number AAH60837.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 1852
Molecular Weight (g/mol) 68.3kDa
Name DUSP9 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 ug
Immunogen MEGLGRSCLWLRRELSPPRPRLLLLDCRSRELYESARIGGALSVALPALLLRRLRRGSLSVRALLPGPPLQPPPPAPVLLYDQGGGRRRRGEAEAEAEEWEAESVLGTLLQKLREEGYLAYYLQGGFSRFQAECPHLCETSLAGRAGSSMAPLPGPVPVVGLGSLCLGSDCSDAESEADRDSMSCGLDSEGATPPPVGLRASFPVQILPNLYLGSARDSANLESLAKLGIRYILNVTPNLPNFFEKNGDFHYKQIPISDHWSQNLSRFFPEAIEFIDEALSQNRGVLVHCLAGVSRSVTVTVAYLMQKLHLSLNDAYDLVKRKKSNISPNFNFMGQLLDFERSLRLEERHSQEQGSGGQASAASNPPSFFTTPTSDGAFELAPT
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias MKP-4/MKP4
Common Name DUSP9
Gene Symbol DUSP9
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.