Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human DYRK2 Synthetic Peptide

Catalog No. PEP1239
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PEP1239 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PEP1239 Supplier Invitrogen™ Supplier No. PEP1239
Add to Cart
Add to Cart

Synthetic Peptide

DYRK2 Synthetic Peptide, PEP-1239, from Invitrogen.

DYRK2 (also known as dual-specificity tyrosine-(Y)-phosphorylation regulated kinase 2) belongs to a family of protein kinases whose members are presumed to be involved in cellular growth and/or development and is defined by structural similarity of their kinase domains and their capability to autophosphorylate on tyrosine residues. DYRK2 expressed in E. coli demonstrated tyrosine autophosphorylation and catalyzed phosphorylation of histones H3 and H2B in vitro. DYRK2 also regulates p53 to induce apoptosis in response to DNA damage.
TRUSTED_SUSTAINABILITY

Specifications

Gene ID (Entrez) 8445
Species Human
Purification Method purified
Accession Number Q92630
Common Name Human DYRK2
Concentration 200 μg/mL
Content And Storage -20°C
Description Synthetic Peptide
Format Liquid
Formulation PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2
For Use With (Application) Blocking Assay, Control
Gene Symbol DYRK2
Lyophilized No
Preservative 0.02% sodium azide
Product Type Peptide
Quantity 50 μg
Regulatory Status RUO - research use only
Storage Buffer PBS
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.