Learn More
Specifications
Specifications
| Assay Range | 0.63 to 20 ng/mL |
| Assay Sensitivity | 0.126 ng/mL |
| Sample Type | Cell Culture Supernatant, Plasma (EDTA), Plasma (Heparin), Saliva, Serum |
| Specificity | Natural and recombinant human E-Cadherin. |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 999 |
| Quantity | 1 Kit |
| Storage Requirements | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
| Synonym | Arc-1, CAD1, cadherin 1, E-cadherin (epithelial), cadherin 1, type 1, E-cadherin (epithelial), cadherin-1, calcium-dependent adhesion protein, epithelial, CAM 120/80, CD324, CD324 antigen, CDH1, CDHE, cell-CAM 120/80, Cell-CAM120/80, ECAD, E-cadherin, ECadherin, Epithelial cadherin, LCAM, L-CAM, UVOE-Cadherin, uvomorulin |
| Target | Human |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
