Learn More
Sino Biological Human EBAG9 PAb 100UL 14012-T56-100
Supplier:  Sino Biological 14012T56100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human EBAG9 (rh EBAG9; Catalog#14012-H07E; O00559; Arg28-Ser213). EBAG9 specific IgG was purified by Human EBAG9 affinity chromatography.
Catalog No. 50-241-1568
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.