Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human EBI2 Alexa Fluor™ 594-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB10272T10
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB10272T10 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB10272T10 Supplier R&D Systems Supplier No. FAB10272T100UG
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

EBI2/GPR183 Monoclonal specifically detects EBI2/GPR183 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen EBI2/GPR183
Applications Flow Cytometry
Classification Monoclonal
Clone 486633
Conjugate Alexa Fluor 594
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Gene Accession No. P32249
Gene Alias EBI2Epstein-Barr virus induced gene 2 (lymphocyte-specific G protein-coupledreceptor), EBV-induced G-protein coupled receptor 2, Epstein-Barr virus-induced G-protein coupled receptor 2, G protein-coupled receptor 183, GPR183, G-protein coupled receptor 183
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived transfected withrecombinant human EBI2, Accession # P32249
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1880
Test Specificity Detects human EBI2 in direct ELISAs.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.