Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human EDA2R/TNFRSF27/XEDAR Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. p-7064161
Change view
Click to view available options
Quantity:
25 μg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB10931SP 25 μg
MAB10931 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB10931SP Supplier R&D Systems Supplier No. MAB10931SP
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

EDA2R/TNFRSF27/XEDAR Monoclonal specifically detects EDA2R/TNFRSF27/XEDAR in Human samples. It is validated for Western Blot, ELISA Capture (Matched Antibody Pair).

Specifications

Antigen EDA2R/TNFRSF27/XEDAR
Applications Western Blot, ELISA
Classification Monoclonal
Clone 173103
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q9HAV5
Gene Alias ectodysplasin A2 receptor, EDA-A2 receptor, EDAA2R, EDA-A2R, EDAA2Rtumor necrosis factor receptor superfamily member XEDAR, EDAR2, TNFRSF27, XEDAR, XEDARTNFRSF27tumor necrosis factor receptor superfamily member 27, X-linked ectodysplasin-A2 receptor
Gene Symbols EDA2R
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human EDA2R/TNFRSF27/XEDAR Met1-Glu136 Accession # Q9HAV5
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 60401
Test Specificity Detects human EDA2R/TNFRSF27/XEDAR in ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human (rh) 41BB, rhBAFF R, rhCD27, rhCD30, rhCD40, rhDR-3, rhDR-6, rhEDAR, rhFAS, rhGITR, rhHVEM, rhLTR-b, rhNGFR, rhOPG, rhOX40 R, rhRANK, rhRELT, rhTAJ, rhTNF R1, rhTNF RII, rhTRADD, rhTRAIL R1, rhTRAIL R2, rhTRAIL R3, rhTRAIL R4, rhTWEAK R, recombinant mouse (rm) DcTRAIL R1, and rmDcTrail R2 is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.