Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human EDAR Biotinylated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. BAF157
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BAF157 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BAF157 Supplier R&D Systems Supplier No. BAF157
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

EDAR Polyclonal specifically detects EDAR in Human samples. It is validated for Western Blot.

Specifications

Antigen EDAR
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Biotin
Dilution Western Blot 0.1 ug/mL
Gene Accession No. Q9UNE0
Gene Alias Anhidrotic ectodysplasin receptor 1, DLED3, Ectodermal dysplasia receptor, ectodysplasin 1, anhidrotic receptor, ectodysplasin A receptor, Ectodysplasin-A receptor, ED1R, ED5, EDA1R, EDA3EDA-A1 receptor, EDA-A1R, Edar, FLJ94390, HRM1, mouse, homolog of, tumor necrosis factor receptor superfamily member EDAR
Gene Symbols EDAR
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human EDAR Glu27-Ile189 Accession # Q9UNE0
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10913
Test Specificity Detects human and mouse EDAR in Western blots. In Western blots, less than 1% cross-reactivity with recombinant human (rh) 4-1BB, rhCD27, rhCD30, rhCD40, rhDR6, rhTAJ, rhTNF RI, rhTNF RII, rhFas, rhGITR, rhHVEM, rhLTR beta , rhNGF R, rhOPG, and rhRANK is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.