Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human EGF-L7 Recombinant Protein, PeproTech®

Catalog No. 10061100UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 μg
100 μg
2 μg
2 x 500 μg
250 μg
50 μg
500 μg
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10061100UG 100 μg
100-61-10UG 10 μg
10-061-01M 2 x 500 μg
10061250UG 250 μg
100-61-2UG 2 μg
10061500UG 500 μg
100-61-50UG 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Catalog No. 10061100UG Supplier Gibco™ Supplier No. 10061100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

100-61-1MG will be provided as 2 x 500 μg (100-61-500UG). Recombinant Human EGF-L7 is a 27.4 kDa protein containing 251 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

Epidermal growth factor-like protein 7 (EGFL7), also known as VE-statin, MEGF7, Notch4-like protein or Zneu1, is a 29.6 kDa protein produced and secreted by endothelium. The protein is also expressed by some normal and malignant tissues. The main function of EGFL7 is to regulate angiogenesis and vasculogenesis. A high level of EFGL-7 can be found in tissues with active angiogenesis of a variety of disorders.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9UHF1
For Use With (Application) Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 51162
Molecular Weight (g/mol) 27.4 kDa
Name Human EGF-L7
Quantity 100 μg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias Cbl20; EGF like domain multiple 7; EGFL7; EGF-like domain 7; EGF-like domain-containing protein 7; EGF-like protein 7; EGF-like-domain, multiple 7; Epidermal growth factor-like protein 7; Estrogen-regulated protein CBL20 20.4 kD; Estrogen-regulated protein CBL20, 20.4 kD; Megf7; multiple EGF-like domain protein 7; multiple EGF-like domains protein 7; multiple epidermal growth factor-like domain protein 7; multiple epidermal growth factor-like domains protein 7; NEU1; NEU1 protein; NOTCH4-like protein; RP11-251M1.2; UNQ187/PRO1449; Vascular endothelial statin; vascular endothelial-statin; VE-STATIN; Zneμ1
Common Name EGFL7
Gene Symbol EGFL7
Conjugate Unconjugated
Recombinant Recombinant
Sequence MYRPGRRVCA VRAHGDPVSE SFVQRVYQPF LTTCDGHRAC STYRTIYRTA YRRSPGLAPA RPRYACCPGW KRTSGLPGAC GAAICQPPCR NGGSCVQPGR CRCPAGWRGD TCQSDVDECS ARRGGCPQRC VNTAGSYWCQ CWEGHSLSAD GTLCVPKGGP PRVAPNPTGV DSAMKEEVQR LQSRVDLLEE KLQLVLAPLH SLASQALEHG LPDPGSLLVH SFQQLGRIDS LSEQISFLEE QLGSCSCKKD S
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.