Learn More
ACROBiosystems Human EGF R 200tests
Supplier:  ACROBiosystems EGRHA2H8200TESTS
Encompass_Preferred
APC-Labeled Human EGF R Protein His Tag (EGR-HA2H8) is produced via conjugation of APC to Human EGF R Protein His Tag with a new generation site-specific technology under Star Staining labeling platform. Human EGF R Protein His Tag is expressed from human 293 cells (HEK293). It contains AA Leu 25 - Ser 645 (Accession # P00533-1).
Catalog No. 50-253-5119
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.