Learn More
Sino Biological Human EGFR MAb 100UL 110463-R0027-100
Supplier:  Sino Biological 110463R0027100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with a synthetic peptide corresponding to the residues around Tyr1068 of Human Phospho-EGF Receptor.
Catalog No. 50-240-9395
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.