Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human EIF2AK3 (NP_004827.3, 536 a.a. - 1116 a.a.) Partial Recombinant Protein with GST-tag

Catalog No. 89942996 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
5 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-942-996 5 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-942-996 Supplier Abnova™ Supplier No. P5619
Add to Cart
Add to Cart

Used for Func, SDS-PAGE

Specifications

Accession Number NP_004827.3
For Use With (Application) Functional Study, SDS-PAGE
Formulation Liquid
Gene ID (Entrez) 9451
Molecular Weight (g/mol) 94kDa
Name EIF2AK3 (Human) Recombinant Protein
Preparation Method Escherichia coli expression system
Purification Method Glutathione sepharose chromatography
Quality Control Testing Loading 1 ug protein in SDS-PAGE
Quantity 5 μg
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias DKFZp781H1925/HRI/PEK/PERK/WRS
Common Name EIF2AK3
Gene Symbol EIF2AK3
Biological Activity The activity was determined by IMAPTM assay. The enzyme was incubated with fluorescein labeled peptide and phosphorylation was detected by IMAPTM technology (fluorescence polarization). Substrate: SRPKtide. ATP: 100μM.
Species E. coli
Recombinant Recombinant
Protein Tag GST
Expression System Escherichia coli expression system
Form Liquid
Purity or Quality Grade 51% by SDS-PAGE/CBB staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.