Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human EIF2D Full-length ORF (NP_008824.2, 1 a.a. - 584 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89943028 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 ug
25 ug
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-943-028 10 ug
89-943-029 25 ug
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-943-028 Supplier Abnova™ Supplier No. H00001939P01S
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

This gene encodes a protein receptor that localizes phosphoglycoproteins within endosomes and at the cell periphery. This trafficking receptor for phosphoglycoproteins may play a role in neuroplasticity by modulating cell-cell interactions, intracellular adhesion, and protein binding at membrane surfaces. In hippocampal neurons, long-lasting down-regulation of ligatin mRNA levels occurs via post-transcriptional RNA processing following glutamate receptor activation. This protein contains single PUA and SUI1 domains and these domains may function in RNA binding and translation initiation, respectively. [provided by RefSeq]

Sequence: MFAKAFRVKSNTAIKGSDRRKLRADVTTAFPTLGTDQVSELVPGKEELNIVKLYAHKGDAVTVYVSGGNPILFELEKNLYPTVYTLWSYPDLLPTFTTWPLVLEKLVGGADLMLPGLVMPPAGLPQVQKGDLCAISLVGNRAPVAIGVAAMSTAEMLTSGLKGRGFSVLHTYQDHLWRSGNKSSPPSIAPLALDSADLSEEKGSVQMDSTLQGDMRHMTLEGEEENGEVHQAREDKSLSEAPEDTSTRGLNQDSTDSKTLQEQMDELLQQCFLHALKCRVKKADLPLLTSTFLGSHMFSCCPEGRQLDIKKSSYKKLSKFLQQMQQEQIIQVKELSKGVESIVAVDWKHPRITSFVIPEPSPTSQTIQEGSREQPYHPPDIKPLYCVPASMTLLFQESGHKKGSFLEGSEVRTIVINYAKKNDLVDADNKNLVRLDPILCDCILEKNEQHTVMKLPWDSLLTRCLEKLQPAYQVTLPGQEPIVKKGRICPIDITLAQRASNKKVTVVRNLEAYGLDPYSVAAILQQRCQASTTVNPAPGAKDSLQVQIQGNQVHHLGWLLLEEYQLPRKHIQGLEKALKPGKKK

Specifications

Accession Number NP_008824.2
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 1939
Molecular Weight (g/mol) 91.1kDa
Name EIF2D (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 ug
Immunogen MFAKAFRVKSNTAIKGSDRRKLRADVTTAFPTLGTDQVSELVPGKEELNIVKLYAHKGDAVTVYVSGGNPILFELEKNLYPTVYTLWSYPDLLPTFTTWPLVLEKLVGGADLMLPGLVMPPAGLPQVQKGDLCAISLVGNRAPVAIGVAAMSTAEMLTSGLKGRGFSVLHTYQDHLWRSGNKSSPPSIAPLALDSADLSEEKGSVQMDSTLQGDMRHMTLEGEEENGEVHQAREDKSLSEAPEDTSTRGLNQDSTDSKTLQEQMDELLQQCFLHALKCRVKKADLPLLTSTFLGSHMFSCCPEGRQLDIKKSSYKKLSKFLQQMQQEQIIQVKELSKGVESIVAVDWKHPRITSFVIPEPSPTSQTIQEGSREQPYHPPDIKPLYCVPASMTLLFQESGHKKGSFLEGSEVRTIVINYAKKNDLVDADNKNLVRLDPILCDCILEKNEQHTVMKLPWDSLLTRCLEKLQPAYQVTLPGQEPIVKKGRICPIDITLAQRASNKKVTVVRNLEAYGLDPYSVAAILQQRCQASTTVNPAPGAKDSLQVQIQGNQVHHLGWLLLEEYQLPRKHIQGLEKALKPGKKK
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias LGTN; HCA56
Common Name EIF2D
Gene Symbol EIF2D
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.